Learn More
METTLER TOLEDO™ SevenExcellence™ S470 pH/Conductivity Benchtop Meter
The S470 meter measures pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET. It can be expanded with additional modules to measure dissolved oxygen, BOD, and ISE. It has an intuitive touchscreen, measurement shortcuts, and can export data to a printer, Excel file, or LIMS system.
Brand: METTLER TOLEDO™ 30046252
2565.00 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET meter than can be expanded with additional measurement modules to measure dissolved oxygen, ISE, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
Specifications
RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA | |
±0.1mV | |
13 Pre defined and 20 user defined standard | |
Max. 5 points, 8 pre defined and 20 user defined buffer group | |
TFT | |
-2000 to 2000 mV | |
0.01 to 100 Mohm/cm | |
0.001 mg/L to 1000 g/L | |
0.001/0.01/0.1 | |
S470 pH/conductivity meter, electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, 10.01, 2 standard sachets each for 1413 uS/cm and 12.88 mS/cm, EasyDirect pH activation key, test certificate | |
0 to 999999mg/L | |
0.1/1mV | |
0.001 to 1 | |
±0.1°C |
±0.5% | |
±0.002 | |
Max. 5 points, 8 pre defined and 20 user defined buffer group | |
20,000 | |
0.001 μS/cm to 2000 mS/cm | |
-2 to 20 | |
0 to 80 ppt | |
0.001 to 1 | |
-30°C to +130°C | |
S470 pH/Conductivity Meter | |
0.001μS/cm to 2000mS/cm | |
0.001 to 1 | |
0.001 to 1 | |
0.1°C |