missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ SmartCal™ Moisture Balance Reference Substances

Verify the performance of your moisture analyzer in just 10 minutes. Mettler Toledo SmartCal Moisture Balance Reference Substances can be run like any other sample to fulfill demands of industry regulation and company audits at the same time.

Brand:  Mettler Toledo™ 30005918

 View more versions of this product

Product Code. 12783275

  • 995.00 € / 12 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Purchase 12 tests, 24 tests, or StarterPac which includes 12 sachets plus measurement protocols, reporting templates, thermo/hygrometer, validation documentation.
This item is not returnable. View return policy

Description

Description

  • Highly sensitive, granular test substance
  • Reliably detects adverse influences from ambient conditions and inaccurate heating or weighing devices
  • Easy to use: pour reference substance into weighing pan, run as typical measurement, review results in 10 minutes
  • Check performance in the QC lab or directly on the factory floor
Test Temperatures
  • Usage range between 70° and 230°C
  • Control limits established for testing at 70°, 100°, 130° and 160°C

 SmartCal™

  • Available in sachets, 12 or 24 per package
  • Supplied with a production certificate issued by Mettler Toledo
cSmartCal™
  • Excellent for highly regulated industries
  • Superior specifications compared to standard SmartCal™
  • Tighter control of instruments
  • Available in sachets, 12 or 24 per package
  • Certified: Supplied by lot with Certificate of Analysis issued by the independent German Federal Institute for Materials Research and Testing in Berlin (BAM, www.bam.de)
StarterPac
  • Recommended for initial SmartCal usage
  • Includes 12 test sachets with all accessories for testing

Order Info

Applicable for testing Mettler Toledo Halogen Moisture Analyzers. Personalized, risk-based testing frequency recommendation is available at www.mt.com/smartcal-frequency-recommendation.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Halogen Moisture Analyzer
12 sachets of cSmartCal plus measurement protocols
12/Pk.
SmartCal StarterPac; 12 Tests Plus Accessories
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.