missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ V20 Karl Fischer™ Titrator 10mL Interchangeable Buret

Interchangeable Buret for use with V20 Karl Fischer Titrator.

Brand:  Mettler Toledo™ 51107501

Product Code. 10136552

  • 2772.96 € / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Rapid titrant change
  • Automatic burette recognition via RFID
  • Serial number on glass cylinder
  • Red protection tubing
  • With PTFE pistons, replaceable valve disks
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Titration Burette
PTFE pistons, replaceable valve disks
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.