Learn More
IBA Lifesciences MEXi-CM
GREENER_CHOICE
IBA Lifesciences GmbH MEXi-CM Culture Medium is an efficient and cost effective mammalian protein expression system.
Brand:  IBA Lifesciences 2-6010-010
Product Code. 15523087
  • 184.00 € / 1000mL
Explore more special offers
Description
Description
MEXi-CM culture medium is a serum-free, chemically defined medium especially developed for growth of high-density MEXi-293E cells. The low biotin content (120 μg/l) enables efficient and convenient purification with the aid of the Strep-tag™ technology.
It is part of the MEXi mammalian expression system. The MEXi system was developed to offer an affordable mammalian protein expression system coming along with all essential components:
• MEXi-293E cell line
• A variety of pDSG expression vectors
• MEXi-CM culture medium
• MEXi-TM transfection medium Further advantages:
• Transient expression for time- and cost-efficient protein production
• Multiple expression vectors with different features
• Optimized for Twin-Strep-tag™:Strep-Tactin™XT system for highly pure proteins
• IP-free system
Specifications
Specifications
For Growth of High-Density MEXi-293 E Cells
1 L