Learn More
Merck MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free
MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free composed of mixed cellulose esters (nitrate and acetate).
Brand: Merck
Description
Composed of mixed cellulose esters (nitrate and acetate)
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications. MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features and Benefits:- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods

Specifications
Not Autoclavable | |
Reusable | |
Disc | |
0.45 μm | |
60 mL/min. x cm2 | |
Plain | |
Hydrophilic | |
13 mm |
White | |
Membrane Filter | |
MCE | |
Non-sterile | |
Clarification of fluids, Sterilizing filtration of tissue culture media, additives, antibiotics, Air monitoring, Particulate analysis, Particulate removal, Biological assays (receptor binding studies, TCA precipitates, etc.). | |
Blotting Membranes | |
0.5 in. |
Safety and Handling
- IMMO-NC triton free MCE 0.45µm WH PL
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose, octylphenol polyethoxyethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.