missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGAT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17869-1-AP-150UL
This item is not returnable.
View return policy
Description
There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. MGAT3 (N-acetylglucosaminyltransferase III) transfers a GlcNAc residue to the beta-linked mannose of the trimannosyl core of N-linked oligosaccharides and produces a bisecting GlcNAc. Expression of this gene may be controlled by a multiple-promoter system.Specifications
MGAT3 | |
Polyclonal | |
Unconjugated | |
MGAT3 | |
GGNT3, GlcNAc T III, GNT III, GNT3, MGAT3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17309, 29582, 4248 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q02527, Q09327, Q10470 | |
Mgat3 | |
MGAT3 Fusion Protein Ag12352 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |