missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Michler's Ketone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 174°C to 176°C |
Boiling Point | >360°C |
Flash Point | 220°C (428°F) |
Quantity | 100 g |
Beilstein | 790733 |
Merck Index | 14,6177 |
Formula Weight | 268.36 |
Percent Purity | 98% |
Chemical Name or Material | Michler's Ketone |
Safety and Handling
Product Identifier
- Michler's ketone
- Danger
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- H318-Causes serious eye damage.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Michlers ketone
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction