missing translation for 'onlineSavingsMsg'
Learn More

Hellma Micro Cuvette, 115F-QS

Highly pure Suprasil™ quartz glass fluorescence cuvette with a PTFE stopper.

Brand:  Hellma 115-F-10-40

Product Code. 10583802

  • 545.00 € / Each
Estimated Shipment: 02-10-2024
to see stock.
Add to basket
This item is not returnable. View return policy

Description

Description

  • High-purity synthetic fused silica quartz construction
  • PTFE stopper
  • For fluorescence measurements
Specifications

Specifications

400 μL
PTFE
PTFE Stopper
10 x 2 mm
Quartz Suprasil
Base thickness: 1.25 mm
1/Pk
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers