Learn More
Hecht Karl™ Micro Hematocrit Capillaries
Brand:  Hecht Karl™ 40563010
Product Code. 10718472
  • 4.00 € / Pack of 100
Explore more special offers
Description
Description
Order Info
LengthMetricTube
Specifications
Specifications
Capillary Tube
Hematocrit Centrifuges
Hematocrit centrifuges
1.1 to 1.2 mm
10Pack
75 mm