Learn More
Fisherbrand™ Micro Insert for well plates, glass
Description
- Clear
- Glass
- Plates meet ANSI and SBS standards
- Different sealing options
- Chromatography tested
- No leeching due to high-end material
Specifications
Clear | |
Glass | |
400 μL | |
Tapered |
Product Content Correction
Product Title
Fisherbrand™ Micro Insert for well plates, glass > clear glass,400 μL Volume,taper bottom
Spot an opportunity for improvement?Share a Content Correction