missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Micro-Ninety™ Cleaning Solution

Use this concentrated cleaning solution for critical applications. Cole-Parmer™ Micro-Ninety™ Cleaning Solution cleans glass, most metals, plastic, rubber, fabric and is safe for all ultrasonic cleaners.

Brand:  Cole-Parmer™ 18100-04


Product Code. 15174626

  • 401.00 € / Pack of 12
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • A mild, yet powerful, free-rinsing cleaner for use in critical cleaning and general purpose applications
  • Radioactive contaminants will not reduce cleaning capability
  • Removes most biological materials
  • Solution works at any concentration and rinses clean; most applications require only 1.25 to 2.5 fluid ounces per gallon of water
  • Solution is environmentally friendly
  • Contains no phosphates, CFCs, ODCs or borates
  • It safely replaces chromic acid to remove protein from glassware
  • Note: Do not use on zinc, magnesium or aluminum 
Specifications

Specifications

Liquid
Yellow
Bottle
9.5
100°C
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • micro 90 concentrated cleaning solution

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : EDTA Na4

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.