missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Micro pH Electrodes

Product Code. p-9397002 Shop All Mettler Toledo Products
Click to view available options
Length (Metric):
40 mm
60 mm
100 mm
130 mm
180 mm
230 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10333243

Brand: METTLER TOLEDO™ 51343160

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

pH electrodes designed for small sample volumes or for measuring in small containers.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. (Connection cable is not included; sold separately.)
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Vials
  • Microplates
  • Test tubes
  • Serum and Gastric juice
  • Micro-biological samples
  • Paint
  • Gel
  • Soap
  • Shampoo
  • Cosmetics

Electrode Specifications

  • InLab Micro: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Micro Pro-ISM: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Ultra-Micro-ISM: ceramic junction; refillable with FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
  • InLab Semi-Micro: ceramic junction; non-refillable - replace when electrolyte depleted
  • InLab Semi-Micro-L: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Flex-Micro: porous PTFE junction; non-refillable - replace when electrolyte depleted

Accessories & Care

  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types S7
Junction Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) Measure pH in the smallest vessels used in the laboratory
For Use With (Equipment) Requires Cable, S7 to BNC (30281915)
Includes ISM Temperature Probe
Internal Reference ARGENTHAL with Ag+ trap
Membrane U Glass
Model InLab™ Micro
Response Time <20 sec.
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 176°F
Length (English) 2.36 in.
Length (Metric) 60 mm
Diameter (English) 0.12 in.
Diameter (Metric) 3 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.