missing translation for 'onlineSavingsMsg'
Learn More

Usbeck™ Micro Spatula Spoon

Mix and transfer substances with simplicity. Usbeck™ Micro Spatula Spoon is ideal for mixing and transferring substances.

Brand:  Usbeck™ 3304

Product Code. 10418191

  • 4.82 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Stainless steel
  • Rigid
  • Width of blade: 5mm
  • Width of spoon 5mm
Specifications

Specifications

Micro Spoon Spatula
Stainless Steel
210 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.