missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell MICRO-TOUCH Nitra-Tex EP
Click to view available options
Size:
Large, 8.5/9
Medium, 7.5/8
Small, 6.5/7
X-Large, 9.5/10
X-Small, 5.5/6
Description
- Additional length for extra protection
- Excellent resistance to a broad range of commonly used chemicals
- Protects from latex Type I allergy
- Tested for use with chemotherapy drugs
Specifications
Specifications
Certifications/Compliance (Europe) | CE 0493, Category III, Medical Device Regulation (EU) 2017/745 Class I, EN 16523-1, EN 421:2010, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 10993-10:2002, ISO 9001, ISO 13485, Food Contact, UKCA |
Chlorinated | Yes |
Accelerator Free | No |
Applications | Healthcare |
AQL | 1.5 |
Color | Blue |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Finger Thickness (English) | 4.7 mil |
Finger Thickness (Metric) | 0.12 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell MICRO-TOUCH Nitra-Tex EP >
Spot an opportunity for improvement?Share a Content Correction