missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL

Brand:  Applied Biosystems™ 403086

1445.85 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10016104

  • 2835.00 € / Pack of 100
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Contains 100 MicroAmp 96-Well Reaction Tube/Tray Retainer Sets. Store at room temperature.

Requires : Caps or full plate covers to seal tubes
This item is not returnable. View return policy

Description

Description

Optimally designed for precise PCR on Applied Biosystems 0.2mL thermal cyclers.

  • Economical 100-unit bulk packs
  • Nonsterile and autoclavable
Greater Sample Uniformity
  • Use with all instruments that have heated sample covers to eliminate condensation and the need for a mineral oil overlay
  • Proprietary MicroAmp Reaction Tubes are essential for high-performance PCR and ensure the greatest temperature and amplification uniformity for samples

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 100 MicroAmp™ 96-Well Reaction Tube⁄Tray Retainer Sets. Store at room temperature.
Natural
2720 Thermal Cycler, 6100 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
100 Assemblies
MicroAmp
96-Well Reaction Tube/Tray/Retainer Set
96-Well Plates
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.