missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL

Brand:  Applied Biosystems™ 403083

 View more versions of this product

Product Code. 10620616

  • 842.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 20 MicroAmp 96-Well Reaction Tube/Tray Retainer Sets.
This item is not returnable. View return policy

Description

Description

Set contains MicroAmpreaction tubes without caps already loaded in a MicroAmp 96-Well Tray/Retainer.

  • Completely pre-assembled for convenience
  • Available in economical 100-unit bulk packs
  • Non-sterile and autoclavable
  • Greater Sample Uniformity
  • Use with all instruments that have heated sample covers to eliminate condensation and need for mineral oil overlay
  • Proprietary MicroAmpreaction tubes are essential for high-performance PCR and ensure greatest temperature and amplification uniformity for samples
  • MicroAmp 96-Well Reaction Tube/Tray/Retainer Set requires caps or full plate covers to seal tubes

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified DNA/RNase Free
MicroAmp
96-Well Reaction Tube/Tray/Retainer Set
PCR
20 Assemblies
Contains 20 MicroAmp™ 96-Well Reaction Tube⁄Tray Retainer Sets. Store at room temperature.
Natural
2720 Thermal Cycler, 7000 System, AB 2720 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
96-Well Plates
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.