Learn More
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plates with Barcode, clear
The Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plate is optimally designed for use with the Applied Biosystems™ Automated Thermal Cycler (ATC).
342.00 € - 2855.00 €
Specifications
Barcode | Includes Barcode |
---|---|
Certifications/Compliance | Certified DNA/RNase Free |
Content And Storage | Store at room temperature. |
Product Line | MicroAmp |
Color | Clear, Optical |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942721
|
Applied Biosystems™
A31728 |
50 Plates |
342.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15990333
|
Applied Biosystems™
A31732 |
500 Plates |
2855.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
These full-skirted reaction plates are manufactured using a durable polycarbonate frame, making them ideal for easy handling with robotic arms and other automated system features. The thin-wall construction of the plate allows for efficient heat transfer and optimal fit in thermal cycler blocks. The black alpha- numeric labeling provides easy-to-read well identification during sample preparation. The MicroAmp EnduraPlate Optical 96-Well Full-Skirted Plate is the only 96-well plate recommended for use with the Automated Thermal Cycler (ATC) in the automated PCR workflow process.
Key features include:
• Designed exclusively for use with Automated Thermal Cycler (ATC) (Cat No. A31486)
• Full-skirted polycarbonate frame for easy robotic handling and plate stacking/loading
• 3-way barcodes on alpha, opposite alpha, and opposite numeric sides to improve plate reading efficiency
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and automated work processes
• Certified free of DNA, RNase, and PCR inhibitors
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete automated workflow needs
Specifications
Includes Barcode | |
Store at room temperature. | |
Clear, Optical | |
Thermal Cycler, Genetic Analyzer, DNA Analyzer | |
Polycarbonate, Polypropylene | |
96 | |
0.2 mL |
Certified DNA/RNase Free | |
MicroAmp | |
Microplate | |
Skirted | |
0.2 mL | |
0.1 mL | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.