Learn More
3M™ Microbial Luminescence System Injector Cleaning Kit GH620522587
Used for routine cleaning of the 3M™ Microbial Luminescence System II (MLS II)
Brand: 3M™ 7000043165
This item is not returnable.
View return policy
Description
- Used with the 3M™ Microbial Luminescence System II (MLS II)
- Removes protein residues and microbial contamination in the injectors and reagent lines, thus avoiding high background readings, false positive results, and inaccurate dispensation of reagents.
Specifications
3 M™ Microbial Luminescence System II (MLS II) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Microbial Luminescence System Injector Cleaning Kit GH620522587
Spot an opportunity for improvement?Share a Content Correction