missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer Microcaps Disposable Glass Micropipettes

Precision-bore glass-capillary tube is topped with a dispensing bulb.

Brand:  Cole Parmer 07950-00

Product Code. 15233495

  • 30.85 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Precision-bore glass-capillary tube is topped with a dispensing bulb.

  • Capillary action moves the fluid up the tube.
  • Squeeze the bulb to release fluid.
  • Accurate to ±1% of rated volume.
  • Each pack contains one bulb unit and 100 disposable tubes.
  • Shipping weight 1 lb. (0.5 kg).
Specifications

Specifications

Glass
0.25 μL
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole Parmer Microcaps Disposable Glass Micropipettes > glass micropipettes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.