Learn More
Cytiva (formerly Pall Lab) Microcheck™ Beverage Monitor
Easy to use, versatile, bulk packaged disposable filter funnels are ideal for microbial analysis.
Brand: Cytiva (formerly Pall Lab) 4763
172.90 EUR valid until 2024-11-29
Use promo code "21718" to get your promotional price.
This item is not returnable.
View return policy
Description
- Can convert to a petri dish for culturing or easily remove the membrane for placing on separate agar dish
- Product not approved for use in medical clinical surgical or other patient applications
Specifications
100 mL | |
Polystyrene Cover, Polypropylene Cylinder/Base, Polyethylene Adapter Material, Cellulose Support Pad | |
50/Pk. | |
Metricel Black Membrane Modified PES |
Black | |
0.45 μm | |
Gamma-irradiated |