Learn More
Cytiva (formerly Pall Lab) Microcheck™ Beverage Monitor with Metricel™ Black Membrane
Easy to use, versatile, bulk packaged disposable filter funnels are ideal for microbial analysis.
Brand:  Cytiva (formerly Pall Lab) 4764
Product Code. 17134401
  • 206.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish
  • Not approved for use in medical clinical surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
50/Pk.
Gridded Filtering Funnel
0.8 μm
Sterile
Metricel Mixed Cellulose Ester