missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Microchem™ 3000 Coveralls with Hood and Integrated Socks
MICROCHEM™ 3000 Coveralls with Hood and Integrated Socks is one of the lightest and most comfortable chemical protective garments on the market today. This durable multi-layer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
663.00 € - 888.00 €
Specifications
Material | Multi-layer Barrier Laminate Fabric |
---|---|
Color | Yellow |
Seam Type | Welded |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15546594
|
Ansell™
YE30W-00122-03 |
Medium |
888.00 €
Pack of 20 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||||
15556594
|
Ansell™
YE30W-00122-04 |
Large |
888.00 €
Pack of 20 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||||
15566594
|
Ansell™
YE30W-00122-05 |
X-Large |
888.00 €
Pack of 20 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
15576594
|
Ansell™
YE30W-00122-06 |
2X-Large |
663.00 €
Pack of 15 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
15586594
|
Ansell™
YE30W-00122-07 |
3X-Large |
663.00 €
Pack of 15 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||||
17101115
|
Ansell™
YE30W-00122-02 |
Small |
888.00 €
Pack of 20 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||||
Description
- Coverall with 2-piece hood
- Integrated socks with boot overflap
- Double zip closure
- Double cuffs
- Elasticated hood, waist, double cuffs
- Excellent liquid penetration resistance and barrier to fine particulates (>0.01 microns)
- Moisture vapour permeable (“breathable”) to help reduce the risk of heat stress
- Silicone free for paint spraying applications
- Low linting to reduces the risk of fibre contamination in some critical areas
- Optimised body fit for wearer comfort and safety
- Anti-static according to EN 1149-5
Specifications
Multi-layer Barrier Laminate Fabric | |
Yellow | |
Welded |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title