Learn More
Ansell™ Microchem™ CFR Coveralls with Hood
MICROCHEM™ CFR is a flame retardant material designed to be worn over woven thermal protective garments such as NOMEX™, offering protection from particulates and pressurised liquid spray without compromising worker protection in the event of a flash fire.
1405.00 € - 1750.00 €
Specifications
Product Type Protective Coveralls
Material Flame-retardant Fabric with PVC Barrier Film
Color Red
Ankle Style Elastic
Cuff Style Elastic
View More Specs
Alert:
Microchem™ CFR should never be worn in isolation for flame retardant protection. Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above.
Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15546484
View Documents
Ansell™
RD96T-00111-03
Medium
1750.00 €
Pack of 25
Estimated Shipment: 02-04-2025
to see stock.
Add to basket
15556484
View Documents
Ansell™
RD96T-00111-04
Large
1750.00 €
Pack of 25
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15566484
View Documents
Ansell™
RD96T-00111-05
X-Large
1750.00 €
Pack of 25
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15586484
View Documents
Ansell™
RD96T-00111-07
3X-Large
1405.00 €
Pack of 20
Estimated Shipment: 02-04-2025
to see stock.
Add to basket
15576484
View Documents
Ansell™
RD96T-00111-06
2X-Large
1405.00 €
Pack of 20
Estimated Shipment: 02-04-2025
to see stock.
Add to basket
Description
Description
  • Coverall with 2 piece hood
  • Elasticated hood, wrists and ankles
  • Double zip closure
  • Flame retardant fabric treated with PVC barrier film
  • Protects from liquid chemicals to EN Type 3 and 4
  • Optimized body fit provides comfort and safety
  • Highly visible bright red colour to improve worker safety
Specifications
Specifications
Protective Coveralls
Red
Elastic
2-panel, Elastic
Stitched, Taped
None
Flame-retardant Fabric with PVC Barrier Film
Elastic
Hooded
Double Zipper
EN14605 Type 3 and 4, EN ISO 13982-1 Type 5, EN 1073-2, EN ISO 14116, EN 1149-5
Includes Hood