Learn More
Thermo Scientific™ MicroClick 24 x 2 Fixed Angle Microtube Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Perform micro-volume applications such as nucleic acid preparation at up to 16,000rpm/ 24,372 x g with the Thermo Scientific™ MicroClick 24 x 2 microtube rotor.
Brand: Thermo Scientific™ 75005715
1375.58 EUR valid until 2024-12-31
Use promo code "21988" to get your promotional price.
This item is not returnable.
View return policy
Description
Perform micro-volume applications such as nucleic acid preparation at up to 17,850rpm/ 30,279 x g with the Thermo Scientific™ MicroClick 24 x 2 microtube rotor.
With Auto-Lock rotor exchange, switch from swing-out to a microtube rotor in as little as 3 seconds with:
- Trouble-free rotor installation and removal
1. No tools are required
2. No complicated rotor changing procedures
- Clear chamber access for cleaning convenience
Biocontainment rotor lid for glove-friendly, one-handed operation, certified by Public Health England, Porton Down, UK.
Ordering Alerts: Replacement biocontainment lid (75005725) and replacement O-rings (75003405) are sold separately. Contact your sales representative for more information.
Specifications
MicroClick 24 x 2 Fixed Angle Microtube Rotor | |
24 x 2 mL | |
16,000 rpm | |
505 | |
11 mm | |
45° |
Fixed Angle Rotor | |
24,372 x g | |
85 mm | |
45 mm | |
Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges | |
Yes |