missing translation for 'onlineSavingsMsg'
Learn More
Bolle Protection™ Microfiber Spectacle Pouch Product Code.: 12619170
Estimated Shipment: 23-04-2025
to see stock.
Add to basket

Bolle Protection™ Microfiber Spectacle Pouch

Product Code. 12619170
Black
Click to view available options
Color:
Black
This item is not returnable. View return policy

Product Code. 12619170

Brand: Bolle Protection™ ETUIFS

Estimated Shipment: 23-04-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Black
Product Type Spectacle Pouch
For Use With (Application) For Spectacles
Model Etuifs
No. per Pack 1 Ea.
Material Microfiber
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bolle Protection™ Microfiber Spectacle Pouch >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.