missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes

Dispense Cassettes for use with MultiFlo FX Dispenser and EL406 Washer Dispenser.
250.00 € - 298.00 €
Specifications
For Use With (Application) | For liquid handling |
---|---|
For Use With (Equipment) | EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser |
Product Code | Brand | Diameter (Metric) | Type | Material | Capacity (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Type | Material | Capacity (Metric) | Price | Quantity & Availability | ||
12335763
![]() |
Agilent BioTek
7170009 |
0.02 cm | Low Volume Replacement Tubing Set | Molded Polypropylene | 1 to 50 μL |
298.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12305763
![]() |
Agilent BioTek
7170008 |
0.035 cm | Medium Volume Replacement Tubing Set | Molded Polypropylene | 5 to 2500 μL |
250.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12365753
![]() |
Agilent BioTek
7170007 |
0.035 cm | Large Volume Replacement Tubing Set | Molded Polypropylene | 10 to 3000 μL |
250.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Volume range 1 μL to 10 mL
- Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
- Dispense 1536 wells in 18 s.
- Cassettes can be autoclaved without recalibration
- Dispense up to eight reagents at one time
- Customizable dispense pattern allows skipped columns or rows
- Built-in priming trough
- Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
- Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput
Specifications
For liquid handling | |
EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes