Learn More
Ansell Microgard™ 1800 Standard Coveralls with Hood
Provides comfort in warmer working environments. Ansell Microgard™ 1800 Standard Coveralls with Hood are constructed of lightweight, breathable material yet protects wearer from light spray and dry particles.
328.00 € - 410.00 €
Specifications
Product Type General Purpose Coveralls
Gender Unisex
Material Microporous Polyethylene Film
Color White
Disposable Single-use
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15516504
View Documents
Ansell
WH18B-00111-02
Small
410.00 €
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15526504
View Documents
Ansell
WH18B-00111-03
Medium
410.00 €
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15536504
View Documents
Ansell
WH18B-00111-04
Large
410.00 €
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15546504
View Documents
Ansell
WH18B-00111-05
X-Large
410.00 €
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15556504
View Documents
Ansell
WH18B-00111-06
2X-Large
328.00 €
Pack of 40
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15566504
View Documents
Ansell
WH18B-00111-07
3X-Large
328.00 €
Pack of 40
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
Description
Description
  • Coverall with 3-piece hood
  • Elasticated wrists, waist and ankles
  • 2-way front zipper with re-sealable storm flap
  • Bound seams
  • Standard: CE category III, type 5/6 EN 1149-5 (antistatic)
  • Low hazard liquid spray, splash and particle protection
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Silicone-free which is critical in spray painting applications
  • Anti-static according to EN1149-5
  • Low linting to reduce the risk of fibre contamination in some critical areas
  • Optimised body fit for wearer comfort and safety
Specifications
Specifications
General Purpose Coveralls
Microporous Polyethylene Film
Single-use
Elastic
Hooded
Front Zipper with Storm Flap
Non-sterile
WH18-B-00-111
Includes Hood
Unisex
White
Elastic
Elastic
3-panel
Bound
CE category III, type 5/6 EN 1149-5
None