Learn More
Ansell Microgard™ 2000 Standard White Coveralls with Hood
Allows perspiration to escape from the suit yet withstands saturation of liquid chemicals. Ansell Microgard™ 2000 Standard White Coveralls with Hood filter 100% of particulates down to 0.01 microns in size.
414.00 €
Specifications
Product Type General Purpose Coveralls
Gender Unisex
Material Microporous Polyethylene Film
Color White
Disposable Single-use
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15536514
View Documents
Ansell
WH20B-00111-02
Small
414.00 €
Pack of 40
Estimated Shipment: 02-04-2025
to see stock.
Add to basket
15546514
View Documents
Ansell
WH20B-00111-03
Medium
414.00 €
Pack of 40
Estimated Shipment: 02-04-2025
to see stock.
Add to basket
15556514
View Documents
Ansell
WH20B-00111-04
Large
414.00 €
Pack of 40
Estimated Shipment: 02-04-2025
to see stock.
Add to basket
15566514
missing translation for 'viewDocuments'
Ansell
WH20B-00111-05
X-Large
414.00 €
Pack of 40
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
15576514
missing translation for 'viewDocuments'
Ansell
WH20B-00111-06
2X-Large
414.00 €
Pack of 40
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
15586514
missing translation for 'viewDocuments'
Ansell
WH20B-00111-07
3X-Large
414.00 €
Pack of 40
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
Description
Description
  • Coverall with 3-piece hood
  • White bound seams
  • Elasticated hood, waist, wrists and ankles
  • Includes fingerloops
  • 2-way front zipper with re-sealable storm flap.
  • Liquid penetration resistance and barrier to fine particulates (>0.01 microns)
  • High quality two-way stretch microporous film provides an effective liquid and particle barrier
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Silicone free - Critical in paint spraying applications
  • Low linting - Reduces the risk of fibre contamination in some critical areas
  • Optimized body fit provides wearer comfort and safety
  • Anti-static treatment
Specifications
Specifications
General Purpose Coveralls
Microporous Polyethylene Film
Single-use
Bound
Hooded
Front Zipper with Storm Flap
Non-sterile
WH20-B-00-111
Includes Hood, Fingerloops
Unisex
White
Elastic
Thumb Loop
3-panel, Elastic
Bound
Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781
None