missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Microgard™ 2000 Ts Plus Coveralls with Hood
Withstands saturation of liquid chemicals and filters 100% of particulates down to 0.01μ. Ansell Microgard™ Ts Plus provides an exceptional overall barrier to low hazard liquid spray and fine particulates.
709.00 €
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Microporous Polyethylene Film |
Color | White |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15526544
|
Ansell
WH20T-00111-02 |
Small |
709.00 €
Pack of 45 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
15536544
|
Ansell
WH20T-00111-03 |
Medium |
709.00 €
Pack of 45 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
15546544
|
Ansell
WH20T-00111-04 |
Large |
709.00 €
Pack of 45 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
15556544
|
Ansell
WH20T-00111-05 |
X-Large |
709.00 €
Pack of 45 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
15576544
|
Ansell
WH20T-00111-07 |
3X-Large |
709.00 €
Pack of 45 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
15566544
|
Ansell
WH20T-00111-06 |
2X-Large |
709.00 €
Pack of 45 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
Description
- Proven barrier to low concentration liquid chemicals, diluted pesticides, liquid and particulate biological hazards
- Moisture vapor permeable to help reduce the risk of heat stress
- Silicone-free for spray painting applications
- Low Linting to reduce the risk of contamination in critical areas
- Anti-static according to EN 1149-1
- Optimized body fit for wearer comfort and safety
- Tunnelled elasticated wrists, hood and ankles minimises the risk of linting and cross contamination
- Thumb loops prevent sleeve movement when working above overhead
- Chinstrap helps to reduce the risk of cross contamination
Specifications
General Purpose Coveralls | |
Microporous Polyethylene Film | |
Single-use | |
Elastic | |
3-panel, Elastic | |
Stitched, Taped | |
EN 14126, EN 1073-2, DIN 32781, EN 1149-1 | |
WH20-T-00-111 | |
Includes Hood with Chinstrap, Thumb Loops |
Unisex | |
White | |
Elastic | |
Hooded | |
Front Zipper with Storm Flap | |
Non-sterile | |
Hood with chinstrap, thumb loops | |
None |