Learn More
Ansell Edmont™ Microgard™ 3000 Cape Hood with Visor Opening
Protects head and shoulders from inorganic chemicals. Ansell Edmont™ Microgard™ 3000 Cape Hood with Visor Opening should also be worn with a protective face shield.
Brand: Ansell Edmont™ YE30W-00508-00
This item is not returnable.
View return policy
Description
- Multi-layer barrier laminate
- Balaclava style with visor to face opening
- Drapes over part of the shoulders
Specifications
Hood with Visor | |
Multi-layer Barrier Laminate Fabric | |
Balaclava style with visor opening | |
30/Pk. |
Universal | |
Yellow | |
Ultrasonically welded |