Learn More
Ansell Edmont™ Microgard™ 3000 Protective Oversleeves Product Code.: 15576604
Estimated Shipment: 05-08-2025
to see stock.
Add to basket
Ansell Edmont™ Microgard™ 3000 Protective Oversleeves
Product Code. 15576604 Shop All Ansell Products
Product Code. 15576604
Brand: Ansell Edmont™ YE30W0060000
Estimated Shipment: 05-08-2025
to see stock.
Add to basket
Add to basket
Fits over the arm just past the elbow to the mid-bicep. Ansell Edmont™ Microgard™ 3000 Protective Oversleeves protect arms from inorganic chemicals.
  • Multi-layer barrier laminate
  • Elasticated at both ends
  • Reaches to mid-bicep
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Product Type Chemical-resistant Sleeve
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Seam Type Ultrasonically Welded
Cuff Style Elastic
Certifications/Compliance Category I PPE according to European Directive 89/686/EEC
Product Content Correction
Product Title
Ansell Edmont™ Microgard™ 3000 Protective Oversleeves >