missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Micrograd™ MICROCHEM™ 5000 Coveralls with Hood
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Coverall with 2-piece hood
- Elasticated double cuffs (knitted inner cuff), waist and ankles
- Seams are ultrasonically welded and taped
- Highly visible bright orange colour for improved worker safety
- Liquid-tight dual zip designs without the need for additional taping
- Provides a barrier to numerous organic and inorganic chemicals and biological hazards
- Comfortable multi-layer material is lightweight, yet strong and durable
- Protection - >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Anti-static according to EN 1149-5
Specifications
Specifications
Product Type | Protective Coveralls |
Size | X-Large |
Material | Multi-layer Fabric |
Color | Orange |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic, Knit |
Neck Style | Hooded |
Hood Type | 2-panel, Elastic |
Closure Type | Liquid-tight Double Zipper |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction