Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
Brand: Epredia™ 524510
105.40 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
- Can be operated from any position on the freeze bar
- Freezing time is shortened if freezing stage and tissue are placed on an active Peltier
Specifications
Heat Extractor for HM560 | |
Heat Extractor | |
HM 550, HM 560 Series |