missing translation for 'onlineSavingsMsg'
Learn More

Linden Micrometer screw gauge in case 0mm to 25mm x 0.01mm

Brand:  Linden H-TOOL-MICROMETER

Micrometer screw gauge in case 0mm to 25mm x 0.01mm

Code : 04

Additional Details : Weight : 0.20000kg

Product Code. 13045903

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers