missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ MicroMop™ Adapters

These lightweight and durable, plastic string mop adapters work with a range of string mops and are available with the choice between a stainless steel handle attachment or an included plastic handle.

54.00 €

Specifications

Product Type Attached Plastic Handle
Color Orange
Length (English) 60 in.
Sterility Non-sterile
Length (Metric) 152 cm
View More Specs

Products 1
Product Code Brand Product Type Material Autoclavable Price Quantity & Availability  
Product Code Brand Product Type Material Autoclavable Price Quantity & Availability  
12827033
View Documents
Micronova™
MH-1
Attached Plastic Handle Fiberglass, Polypropylene Autoclavable
54.00 €
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
Description

Description

The Micronova MicroMop™ String Mop Adapters work with a variety of string mops, large and small, and are available with a choice of two handle options. The QDMH-1 adapter, with a Quick Disconnect handle attachment, connects to the stainless steel SSU series handles. The MH-1 adapter includes an attached, 60-inch plastic handle.

  • Orange polypropylene string mop holders
  • Lightweight and durable
  • Holds a range of string mops
  • QDMH-1 Adapter features a Stainless Steel, Quick Disconnect Handle Attachment
  • MH-1 Adapter includes an attached 60-inch plastic coated handle
  • Autoclavable
Specifications

Specifications

Attached Plastic Handle
60 in.
152 cm
Reusable
MicroMop™ and PolyGen™ Series Mops
Fiberglass, Polypropylene
Orange
Non-sterile
Autoclavable
Store at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
For Cleaning Large Surface Areas Quickly and Efficiently
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.