missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Isolator Tool and Irradiated Covers
BIOPHARMAGREENER_CHOICE

The irradiated angled head adapter with a 360° swivel works with a range of irradiated mop covers to efficiently clean and disinfect isolators, hoods, RABS and small equipment in clean and aseptic environments.

82.35 € - 665.00 €

Specifications

Disposable Reusable
View More Specs

Products 6
Product Code Brand Product Type Material Price Quantity & Availability  
Product Code Brand Product Type Material Price Quantity & Availability  
13137666
View Documents
Micronova™
QPIT-09
Plastic Isolator Tool, ACME Threaded Handle Attachment Acetal
82.35 €
Each
Estimated Shipment: 15-07-2025
to see stock.
Add to basket
 
13128675
View Documents
Micronova™
ITC620-9IR
Isolator Cleaning Tool Cover Polyester This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13118675
View Documents
Micronova™
ITC620-911IR
Isolator Cleaning Tool Cover Non-woven Polyamide
665.00 €
Pack of 50
Estimated Shipment: 31-07-2025
to see stock.
Add to basket
 
12817033
BIOPHARMAGREENER_CHOICE
View Documents
Micronova™
ITC6-9IR
Isolator Cleaning Tool Cover Elastic Band
509.00 €
Pack of 50
Estimated Shipment: 15-07-2025
to see stock.
Add to basket
 
15541755
BIOPHARMAGREENER_CHOICE
View Documents
Micronova™
ITC11-9NSIR
Isolator Cleaning Tool Cover Textured Polyester
414.00 €
Pack of 50
Estimated Shipment: 15-07-2025
to see stock.
Add to basket
 
11932485
View Documents
Micronova™
QDIT-09
Isolator Tool, Quick Disconnect Handle Attachment Stainless Steel (304)
182.00 €
Each
Estimated Shipment: 15-07-2025
to see stock.
Add to basket
 
Description

Description

Help to simplify cleaning in compliance with USP 797

The Micronova Irradiated Isolator Tool is designed for use with the SSU Series Universal Handles or STA Series threaded handles to fit through small passes and allow extended reach. The mop covers are available in a range of fabrics with optional foam layer to hold disinfectant and facilitate wringing.

  • Irradiated plastic isolator tool with choice of irradiated covers
  • Ergonomic cleaning for isolators, hoods, RABS and other small equipment
  • Modular and extendable handles fit through small passes and expand reach
  • 360° swivel, angled head adapter for easy application of cleaning agent
  • Gamma irradiated for use in aseptic environments
  • Covers available in a variety of fabrics for specialized cleaning
    • NovaPoly — 100% smooth knit polyester provides excellent disinfectant dispersal
    • PolySorb — 100% textured polyester for removing residues and dispersing disinfectant
    • MegaTex — 100% non-woven polyamide is absorbent with a mildly abrasive surface
  • Compatible with alcohol, strong disinfectants and steam sterilization
  • Launderable for reuse
  • Autoclavable
Specifications

Specifications

Reusable
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.