Learn More
Micronova™ Hand Held Scraper
A hand held, polypropylene scraper tool with an injection-molded, seamless design that prevents bacterial growth during residue and build-up removal in controlled environments.
Brand:  Micronova™ HHSWH-8
 View more versions of this product
Product Code. 13127286
  • 25.25 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Micronova Hand Held Scraper features injection-molded, polypropylene construction, without seams or cracks that might harbor bacterial growth. The scraper is autoclavable and will not scratch metal surfaces.
  • Injection-molded, hand held scraper tool
  • For removing residue and build up in controlled environments
  • White polypropylene construction
  • No seams or cracks to harbor bacterial growth
  • Will not scratch metal surfaces
  • Available in 8 or 10-inch lengths
  • Autoclavable
Specifications
Specifications
Scraper Tool
8 in.
20.3 cm
7.6 cm
Chemicals
Reusable
Store at Ambient Temperature
FDA Compliant
White
Non-sterile
Autoclavable
3 in.
7.62 x 20.32 (3 x 8 in.)
1/Pk., 12/Cs.
Controlled Environment Residue Removal
Polypropylene