Learn More
Micronova™ Irradiated MegaSwat™ Mop Covers
Micronova™'s irradiated, mop covers work with the blade style MegaSwat™ Mop adapter to effectively clean around, under and behind heavy processing equipment and calibrated apparatus in the lab and cleanroom.
263.00 € - 385.00 €
Specifications
Care Instructions | Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C. |
---|
Product Code | Brand | Dimensions | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
13177436
![]() |
Micronova™
MS6-8IR |
7.6 x 20.3 cm (3 x 8 in.) | Polyester | QDMS-16 and SSU Series Handle |
263.00 €
Pack of 12 |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||
13187426
![]() |
Micronova™
MS611-16LIR |
7.6 x 40.6 cm (3 x 16 in.) | Smooth Knit Polyester, Urethane Foam | QDMS-16 and SSU Series Handle |
385.00 €
Pack of 12 |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||
Description
The Micronova™ Irradiated MegaSwat™ Mop Covers are designed to work with the MegaSwat™ Mop Adapter and SSU series QD handles, for wet or dry cleaning and disinfecting in ISO 5 to 9 areas. The flexible blade style mop is ideal for cleaning under, around and behind heavy or calibrated equipment in the cleanroom. The covers are gamma irradiated and available with or without a foam interior.
- Knit polyester mops easily glide under and behind equipment
- Designed for use with flexible MegaSwat™ Mop Adapter and SSU series QD handles
- Available in various lengths
- Available with or without foam interiors
- NovaPoly, 100% polyester knit provides excellent disinfectant dispersal
- Compatible with alcohol and strong disinfectants
- Launderable for reuse
- Autoclavable
Specifications
Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C. |