Learn More
Micronova™ MegaTape™ Polyethylene Cleanroom Tape
A medium density, polyethylene tape with hi-tack/clean-peel acrylic adhesive, good conformability and excellent resistance to moisture, oils and weathering. Ideal for clean construction and maintenance projects.
11.90 € - 18.55 €
Specifications
Color White
Material Polyethylene
Sterility Non-sterile
Length (English) 36 yd.
Length (Metric) 33 m
View More Specs
Products 2
Product Code Brand Width (Metric) Width (English) Price Quantity & Availability  
Product Code Brand Width (Metric) Width (English) Price Quantity & Availability  
13167326
View Documents
Micronova™
MTP-2WH
50.8 mm 2 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13157326
View Documents
Micronova™
MTP-1WH
25.4 mm 1 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
The Micronova MegaTape™ Polyethylene Cleanroom Tape, with hi-tack acrylic adhesive, provides superior sealing abilities and removes cleanly with no adhesive residue. This tape is low outgassing, has good conformability, similar to vinyl tapes, and shows excellent resistance to UV, moisture, acids, alkalis, oils, abrasion and weathering.
Specifications
Specifications
White
Non-sterile
33 m
Polyethylene
36 yd.