Learn More
Micronova™ PharmaPly™ Mop Head
BIOPHARMA
Ideal for covering large surface areas quickly and efficiently.
Brand:  Micronova™ PPM4-MM15
Product Code. 11906875
  • 442.00 € / Pack of 24
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyester/rayon blend mop head for cleaning/disinfecting pharmaceutical support areas, hospitals and laboratories
  • Designed for use with MicroMop™ adapter and handle (sold separately)
  • 15 in. long (38cm)
Specifications
Specifications
Cleanroom Mop
15 in.
Autoclavable
Reusable
No
White
Sterile
6 in.
Used for Covering Large Surface Areas Quickly and Efficiently, Floor
Polyester, Rayon