Learn More
Micronova™ PolyMitt™ Cleaning Mitt Wipe
A range of cleaning mitts that offer the versatility of a high quality wipe with the convenience and security of a glove for simplified cleaning around hard to reach areas in the cleanroom. Available in a variety of fabrics for specialized cleaning.
6.85 € - 14.35 €
Specifications
Length (English) | 12 in. |
---|---|
Length (Metric) | 30.5 cm |
Width (English) | 8 in. |
Width (Metric) | 20.3 cm |
Disposable | Reusable |
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
13187746
|
Micronova™
MW-AA |
Blue | Polyester/Nylon Microfiber |
8.30 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||||
13157756
|
Micronova™
MWDT-AA11 |
Blue | Polyester/Nylon Microfiber, Double-sided Thin Urethane Foam | N/A | |||||
13147746
|
Micronova™
MW-6 |
White | Smooth-knit Polyester | N/A | |||||
13187736
|
Micronova™
MW-2011 |
White | Non-woven Polyamide, Urethane Foam |
11.15 €
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
13167736
|
Micronova™
MW-11 |
White | Textured Polyester | N/A | |||||
Description
The Micronova PolyMitt™ Cleaning Mitt Wipes come in one standard size, designed to fit comfortably over a gloved hand, making it easier to clean around carts, shelving, tanks, tubing and other hard to clean areas in the cleanroom.
- Durable cleaning mitts for simplified cleaning
- Versatility of a high quality wipe with the convenience and security of a glove
- One size: fits comfortably over a gloved hand
- Designed to clean hard to reach areas in the cleanroom
- Available in a range of fabrics for specialized cleaning:
- NovaPoly — 100% smooth knit polyester provides excellent disinfectant dispersal
- PolySorb — 100% textured polyester for removing residues and dispersing disinfectant
- MegaTex — 100% non-woven polyamide is absorbent with a mildly abrasive surface
- Microfiber — 80% polyester, 20% polyamide is highly absorbent and effective wet or dry
- Available with or without a urethane foam layer
- Compatible with strong disinfectants
- Autoclavable
- Reusable
Specifications
12 in. | |
8 in. | |
Reusable | |
Cleaning Mitt for Cleanroom and Controlled Environment Surfaces | |
Non-sterile |
30.5 cm | |
20.3 cm | |
30.5 x 20.3 cm (12 x 8 in.) | |
1/Pk., 24/Cs. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.