missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Stainless Steel Quick Disconnect SSU Handles

The fundamental electropolished stainless steel handles with quick disconnect attachments that work with all Micronova QD series mop adapters and cleaning tools. Available in a range of fixed and extendible lengths.

55.60 € - 383.00 €

Specifications

Sterility Non-sterile
Autoclavable Autoclavable
Resistant To Chemicals, Corrosion
Care Instructions Store at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
For Use With (Equipment) Mops
View More Specs

Products 12
Product Code Brand Product Type Length (English) Length (Metric) Price Quantity & Availability  
Product Code Brand Product Type Length (English) Length (Metric) Price Quantity & Availability  
13177506
View Documents
Micronova™
SSU-8
Extendible Handle 14 to 23 in. 35.5 to 58 cm
176.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
13127506
View Documents
Micronova™
SSU-1836
Extendible Handle, Quick Disconnect Attachment 18 to 36 in. 46 to 91 cm
213.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
11976845
View Documents
Micronova™
SSU-1
Extendible Handle, Quick Disconnect Attachment 38 to 72 in. 96.5 to 183 cm
310.00 €
Each
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
 
17501725
View Documents
Micronova™
SSU-10
Extendible Handle 47 to 80 in. 119 to 203 cm
252.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11986845
View Documents
Micronova™
SSU-3
Extendible Handle, Quick Disconnect Attachment 62 to 120 in. 157 to 305 cm
383.00 €
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
 
13187506
View Documents
Micronova™
SSU-9
Fixed Handle 12 in. 30.5 cm
55.60 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
13167506
View Documents
Micronova™
SSU-6
Fixed Handle 25 in. 63.5 cm
58.95 €
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
 
13157506
View Documents
Micronova™
SSU-5
Fixed Handle, Quick Disconnect Attachment 37 in. 94 cm
70.40 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
12837033
View Documents
Micronova™
SSU-4
Fixed Handle, Quick Disconnect Attachment 61 in. 155 cm
104.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
13147506
View Documents
Micronova™
SSU-2
Fixed Handle
71.05 €
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
 
11996845
View Documents
Micronova™
SSU-7
Fixed Handle 51 in. 130 cm
101.00 €
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
 
13137506
View Documents
Micronova™
SSU-1S
Universal Extendible Handle 38 to 72 in. 96.5 to 183 cm
226.00 €
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
 
Description

Description

Entire handle can be autoclaved

The Micronova Stainless Steel Quick Disconnect SSU Handles are made from 304 electropolished stainless steel with polyketone components that provide strong, hard-wearing all-around connections for efficient and long lasting use. The included Quick Disconnect attachment is designed for use with the QD series mop adapters and cleaning tools.

  • Fundamental handles that work with all Micronova mopping systems
  • Electropolished stainless steel with Polyketone components
  • Strong, hard-wearing connections for long lasting use
  • Available fixed or extendible:
    • Fixed length handles available from 8 in (20 cm) to 5 feet (1.5 m)
    • Extension handles available from 3 feet (1 m) to 10 feet (3 m)
  • For use with all QD series adapters
  • May be used with QP tools and frames with addition of the SSA adapter
  • Good compatibility with disinfectants
  • Autoclavable
Specifications

Specifications

Non-sterile
Chemicals, Corrosion
Mops
Autoclavable
Store at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
Mop Handle for Cleaning and Sanitizing Cleanrooms
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.