Learn More
Socorex™ Micropipette Rack for Qualitix™ Tipfill Refill System
Reusable, reclosable, and more space-saving than conventional racks. Socorex™ Micropipette Rack for Qualitix™ Tipfill Refill System readily accepts micropipette refill packs and has smooth, rounded edges and an ergonomic snap latch that won't catch gloves.
Brand:  Socorex™ 3070.1000ER
 View more versions of this product
Product Code. 15790649
  • 30.85 € / Pack of 6
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accepts Qualitix™ Tipfill Refill System refill packs.
  • Hinged lid.
  • Smooth, rounded edges.
  • Ergonomic snap latch that won't catch gloves.
  • Transparent lid.
  • Reusable.
Specifications
Specifications
Yes
Autoclavable
Micropipette Tips (1000μL)
Qualitix™Tipfill Refill System
6
96
Natural
Yes
8