missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microplate Shakers with 4-Place Platform

For life science, immunoassay. shaking ELISA plates, staining cells for flow cytometry,shaking plates in cold room, food and beverage, histamine in cheese, vitamins in milk testing applications

830.00 €

Specifications

Orbit 2.5 mm
Load Bearing Capacity (Metric) 5 kg
Speed Control Accuracy Speed Accuracy: ±1 rpm
Speed Range 150 to 1000 rpm
Voltage 100/240 V
View More Specs

Products 1
Product Code Brand Plug Type Price Quantity & Availability  
Product Code Brand Plug Type Price Quantity & Availability  
15504070
View accessory products View Documents
Fisherbrand™
88861024
EU, UK, CN Plug
830.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Variable shaking speed: 150 to 1000rpm 
  • Quick and easy fitting or any standard-depth microplates 
  • Continuous or timed operation, with automatic switch off 
  • Display will show last used settings, even after power has been turned off.
  • Continue to run by last set speed and the remaining time when the power recovered.
  • Operating temperature 5°C to 40°C allows for use in refrigerators, cold rooms and non-CO2 incubators
  • Holds up to four microplates  
  • Easy-to-use electronic timer ensures accurate count-down and repeatability of time sensitive incubations
Specifications

Specifications

2.5 mm
Speed Accuracy: ±1 rpm
100/240 V
Aluminum Platform
415 x 293 x 143mm
0 min. to 99:59 hr.
5 kg
150 to 1000 rpm
5°C to 40°C
320 x 260 mm
LED
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.