Learn More
Invitrogen™ Microplates for Fluorescence-based Assays
Microplates for Fluorescence-based Assays are black-walled, clear-bottom 384-well microplates that minimize well-to-well crosstalk and autofluorescence. These microplates are optimized for fluorescence-based applications including nucleic acid quantitation (e.g. Quant-iT assays).
Brand:  Invitrogen™ M33089
174.80 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
 View more versions of this product
Product Code. 15329413
  • 184.00 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Molecular Probes® 96-well microplates for fluorescence-based assays are black-walled, clear-bottom 96-well microplates that minimize well-to-well crosstalk and autofluorescence and so are optimized for fluorescence-based applications. These plates are compatible with standard fluorescence microplate readers and are offered in a convenient 10-pack size for those times when only a few assay plates are required.
  • Black-walled, clear-bottom 96-well microplates that minimize well-to-well crosstalk and autofluorescence
  • Compatible with standard fluorescence microplate readers
  • Packaged in a convenient 10-pack size
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at room temperature.
Nunc
96
10 plates
10-plates
Microplate Reader
Non-sterile