Learn More
Berkshire™ MicroPolx™ 2750 Microdenier Wiper
MicroPolx™ 2750 is a cleanroom laundered polyester/nylon microfiber knitted wiper.
120.00 €
Specifications
Product Type | Flat Head Mop Cover |
---|---|
Sterility | Non-sterile |
Material | Polyester, Synthetic Microfiber |
Description
- Ultrafine microfiber yarn increases surface area and provides superior wiping efficiency
- Wedge shaped fiber cross section lifts, entraps and removes particulates from critical surfaces
- Cleans efficiently with less solvent usage than traditional wipers
- Removes oily soils with little or no solvent required
- Soft hand and non-abrasive ultrasonic sealed edge are excellent for sensitive or scratch prone surfaces
- Continuous filament yarns and tight knit construction provide excellent abrasion resistance and low fiber shedding
- Chemically compatible with IPA and other commonly used solvents
Specifications
Flat Head Mop Cover | |
Non-sterile | |
Polyester, Synthetic Microfiber |