missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Waterproof Portable pH/pH-mV/ORP/Temperature Meter with Sensor Check™

Allows the user to determine the electrode status at any time. Hanna Instruments™ Waterproof Portable pH/pH-mV/ORP/Temperature Meter with Sensor Check™ is supplied with a rugged carrying case and a unique titanium body pH/ORP electrode that has a built-in temperature sensor.

Brand:  Hanna Instruments™ HI991003

Product Code. 11667260

  • 1700.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This meter features a large multi level LCD screen which displays both pH/pH-mV/ORP and temperature readings simultaneously with 0.01 pH resolution and an accuracy of ±0.05 pH. The LCD screen also has indicators for calibration status and stability, as well as on-screen tutorial messages. The HI991003 has many advanced features that are found in more expensive portable instrumentation including Sensor Check, automatic calibration, buffer recognition,and temperature compensation. The battery percent level is displayed at start up alerting the user to the remaining battery power that is available.

  • Waterproof: Designed to float if accidentally dropped in a tank
  • Sensor Check™: Allows users to check the electrode status at any time
  • Expanded pH and ORP Range: pH from -2.00 to 16.00, ORP is ±1999 mV
  • Automatic Two-Point Calibration: One or two-point calibration is automatic to two selectable buffer sets (4.01, 7.01 and 10.01 or NIST 4.01, 6.86, and 9.18)
  • Automatic Temperature Compensation
  • Temperature displayed in °C or °F along with pH reading
  • Stability Indicator: Clock tag will disappear when the reading has achieved stability
  • HOLD button: Freezes reading on the display to allow recording of measurement
  • Exceptional long battery life of 1200 hours of continuous use
  • BEPS (Battery Error Prevention System): Meter will automatically shut off if there is not enough power to get an accurate measurement
  • Battery % level at startup and Low Battery Indicator (below 5%)
  • Auto-Off: Meter automatically shuts off after 8 minutes of non-use to maximize battery life
Specifications

Specifications

HI1297D pH/ORP probe with internal temperature sensor, HI70004 pH 4.01 buffer sachet (20 mL), HI70007 pH 7.01 buffer sachet (20 mL), HI700661 electrode cleaning solution sachet (2 x 20 mL), batteries, instructions, and rugged carrying case
±2
Automatic
0.01
1mV
-5°C to +105°C
0.1°C, 0.1°F
152 x 58 x 30 mm (6.0 x 2.3 x 1.2 in.)
7.2 oz.
HI1297D pre-amplified pH/ORP probe with internal temperature sensor, DIN connector and 1m cable
automatic from -5.0 to 105.0°C (23 to 221°F)
0°C to 50°C
Ph + Temperature Meter
58 mm
±0.5°C (up to 60°C); ±1.0°C (outside), ±1°F (up to 140°F);
±0.02
-2.00 to 16.00
±1999
1 or 2 with 2 sets of standard buffers (pH 4.01, 7.01, 10.01 or pH 4.01, 6.86, 9.18)
23°F to 221°F
LCD
205 g
2 Years (6 Months Probe)
1
32°F to 122°F
3 x 1.5V AAA
1200 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.