Learn More
Sartorius Microsart™ ADDfilter, White-Black, High Flow
Ready-to-use, sterile Microsart™ ADDfilter High Flow units combine a funnel, filter base and gridded membrane filter in one unit. They have been specifically developed for the detection and enumeration of microorganisms in pharmaceuticals, cosmetics, food, beverages, water and other liquids.
Marke: Sartorius 16D01--25-H6--BS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- The Microsart™ ADDfilter range has been tailored to meet individual needs.
- There are two volume sizes to choose from, 100 mL and 250 mL, different pore sizes and different filter colors for contrasting backgrounds during evaluation.
- Marked graduations allow accurate sample volumes
- The optimal design permits a thorough rinsing of the system following filtration.
- No liquid is retained in the filter funnel.
- Optimal sealing to the Microsart™ Base 47 mm is guaranteed by the Click-Fit closure.
- The filter units are available as tray versions with lids, or are stacked in bags for safe removal using the Microsart™ Funnel Dispenser.
- Microsart™ ADDvance is an entire product family and stands for innovative products for microbiological analyses based on colony count determination.
- From taking the sample until incubation in just a few easy steps, whilst at the same time minimizing the risk of secondary contamination.
- All Microsart™ products meet the most stringent quality assurance standards, providing reliable results and offering simple, time-saving handling.
- Microsart™ ADDvance is advanced colony counting.
- Microsart™ Base and Funnel Dispenser
- Microsart™ Base 47 mm is the perfect addition to existing Combisart™ and Microsart™ Combi.jet stainless steel manifolds.
- The Microsart™ Funnel Dispenser guarantees secure removal of single, sterile Microsart™ ADDfilter units.
- Even after opening the bag, the remaining funnels are protected from secondary contamination.
- The pharmaceutical and biotech industries are driven by the need to optimize their work flows and increase efficiency without compromising their level of safety.
- Products and raw materials used in the pharmaceutical or biotech industry require control of microbial levels during processing and handling.
- All components of the filtration system must comply with international guidelines, such as USP, EP or ISO standards.
Spezifikation
Cellulose Nitrate | |
250 mL | |
216 Pk. |
0.45 μm | |
Sterile | |
Sterile |