missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ MicroSeal SuperSorb™ Wipers
BIOPHARMA

Sealed Edge Cleanroom Laundered 100% Polyester Knit wiper. ISO Class 3 to 5 (Cleanroom Class 1 to 100).

Brand:  Berkshire™ MSSS.0909.8

 View more versions of this product

Product Code. 15526344

  • 1215.00 € / Pack of 800
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 100% continuous filament polyester knit
  • Two-ply pinsonic construction with unique knit structure and pattern
  • Ultrasonically sealed edge for reduced fiber contamination
  • Laundered and packaged in Berkshire’s ISO Class 4 cleanroom
Specifications

Specifications

Cleanroom Wipe
22.86 cm
22.86 cm
IPA, Acetone and other solvents
RoHS 2002/05, REACH
Sealed
Chamber cleaning and cmP processing
Non-sterile
9 in.
9 in.
Polyester Knit
267
White
9 x 9 in. (22.86 x 22.86 cm)
2-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers