missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Microsep™ Advance Centrifugal Devices with Supor Membrane

Particulate removal for longer HPLC and UHPLC column life.

Brand:  Cytiva (formerly Pall Lab) MCPM45C68

 View more versions of this product

Product Code. 17194751

  • 895.00 € / Pack of 100
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High spin speed and larger EFA reduces spin times
  • For samples from 0.5 to 5 mL
  • For research applications only
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wildberry
PES
Non-sterile
0.5 to 5 mL (Sample)
EFA: 3.3 cm2
0.45 μm
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers